Assay Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300933.1 | NP_001287862.1 | 5 | 690 | 71 | |
| NM_001300934.1 | NP_001287863.1 | 3 | 525 | 71 | ||
| NM_001300935.1 | NP_001287864.1 | 5 | 723 | 71 | ||
| NM_207172.1 | NP_997055.1 | 5 | 677 | 71 | ||
| NM_207173.1 | NP_997056.1 | 5 | 677 | 71 | ||
| GenBank mRNA | AY255536.1 | - | 4 | 153 | 71 | |
| AY310326.1 | - | 5 | 677 | 71 | ||
| AY310327.1 | - | 5 | 677 | 71 | ||
| AY310328.1 | - | 5 | 644 | 71 | ||
| AY310330.1 | - | 4 | 573 | 71 | ||
| AY310331.1 | - | 4 | 583 | 71 | ||
| AY310332.1 | - | 3 | 479 | 71 | ||
| AY956446.1 | - | 5 | 582 | 71 | ||
| AY956447.1 | - | 5 | 553 | 71 | ||
| AY956448.1 | - | 5 | 553 | 71 | ||
| BC132693.1 | - | 5 | 594 | 40146715 | 71 | |
| BC132695.1 | - | 5 | 594 | 40146717 | 71 | |
| BC144380.1 | - | 3 | 396 | 9052901 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NPSR1
Genomic Map