Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002064.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002064.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002064.hg.1 |
TC11000143.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100147257.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147257.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147257.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147257.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147257.hg.1 |
TC1100006750.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
ENST00000299481.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006750.hg.1 |
NM_176822.3 |
Refseq |