Assay Details
| Gene Symbol: SPNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320449.1 | NP_001307378.1 | 1 - 2 | 238 | 89 | |
| NM_182538.4 | NP_872344.3 | 1 - 2 | 238 | 89 | ||
| XM_011523723.1 | XP_011522025.1 | 1 - 2 | 222 | 89 | ||
| XM_011523724.1 | XP_011522026.1 | 1 - 2 | 222 | 89 | ||
| XM_011523725.1 | XP_011522027.1 | 1 - 2 | 222 | 89 | ||
| GenBank mRNA | AK172832.1 | - | 1 - 2 | 238 | 89 | |
| BC023646.2 | - | 1 - 2 | 190 | 5087991 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000041.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC023646.2,NM_001320449.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17000411.hg.1 | ENST00000333476.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000411.hg.1 | ENST00000355530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000411.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000411.hg.1 | TC17000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000410.hg.1 | ENST00000333476.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000410.hg.1 | ENST00000355530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000410.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000410.hg.1 | TC17000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000409.hg.1 | ENST00000333476.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000409.hg.1 | ENST00000355530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000409.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000409.hg.1 | TC17000041.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000572078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000575796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006626.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145230.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145230.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145230.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145230.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145230.hg.1 | TC1700006626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145234.hg.1 | TC1700006626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000355530.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000575194.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | ENST00000576069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | NM_182538.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700145235.hg.1 | TC1700006626.hg.1 | Clariom D TC |

SPNS3
Genomic Map