| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023323.hg.1 |
TC03001361.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023323.hg.1 |
ENST00000429665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023323.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023325.hg.1 |
TC03001361.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023325.hg.1 |
ENST00000429665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023325.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023322.hg.1 |
TC03001361.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023322.hg.1 |
ENST00000429665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023322.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023322.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179976.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179976.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179976.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179976.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179976.hg.1 |
TC0300010960.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179978.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179978.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179978.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179978.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179978.hg.1 |
TC0300010960.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010960.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
ENST00000429665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
ENST00000447340.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
NM_182702.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179975.hg.1 |
TC0300010960.hg.1 |
Clariom D TC |