Assay Details
Gene Symbol: KRTAP22-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181620.1 | NP_853651.1 | 1 | 49 | 88 | |
GenBank mRNA | AB096953.1 | - | 1 | 83 | 88 | |
BC101682.1 | - | 1 | 75 | 8069188 | 88 | |
BC101686.1 | - | 1 | 75 | 8069192 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000105.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005754 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11346265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11346273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11346269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11346281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005752 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005755 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005758 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005743 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32005746 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3407672 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3207 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3208 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P115070 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P115070 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40205 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P21232342 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346268 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005760 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005750 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346263 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005742 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346274 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005749 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005754 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005744 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005757 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346265 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346264 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005748 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346278 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346269 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005758 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346281 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005752 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005759 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005746 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346266 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005761 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005743 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005755 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346273 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005756 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005745 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346275 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346271 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005751 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005747 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005755 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005757 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005745 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346278 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005754 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005752 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346266 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005751 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005759 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346268 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346281 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346271 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346273 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005749 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005750 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005746 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346275 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346263 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005744 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005748 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005756 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346264 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005760 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346277 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346265 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005761 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346269 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005755 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005742 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005758 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P11346274 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005743 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P32005747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005755 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11346273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32005745 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115070 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346277 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346268 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346271 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346272 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346270 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346281 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346275 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11346274 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115070 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P115070 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 114297 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 11563 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346262 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346252 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346245 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346238 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346227 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346225 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346281 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346284 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346249 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346268 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346274 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346283 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346222 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346224 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346271 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346236 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346232 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346256 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346257 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11346229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_181620_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8068153 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8068152 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8068151 | - | - |
Affymetrix Human Genome U219 Array | 11738605_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738605_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR21000664.hg.1 | ENST00000334680.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21000664.hg.1 | NM_181620.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21000664.hg.1 | TC21000105.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100006879.hg.1 | NM_181620.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146336.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146336.hg.1 | NM_181620.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146336.hg.1 | TC2100006879.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100146336.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146336.hg.1 | NM_181620.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146337.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146337.hg.1 | TC2100006879.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100146337.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006879.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100006879.hg.1 | NM_181620.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100146335.hg.1 | ENST00000334680.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100146335.hg.1 | TC2100006879.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100146335.hg.1 | ENST00000334680.3 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019141 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019141 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 516344 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1678828 | BC101682.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1678828 | BC101686.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31791011-S | BC101682.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31791011-S | BC101686.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31791011-S | NM_181620.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31791011-S | AB096953.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_24494-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003610563 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003610563 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 870100 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_24494 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1678828 | AB096953.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1678828 | BC101682.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1678828 | BC101686.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1678828 | NM_181620.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1678828 | AB096953.1 | human_transcriptome |
