Assay Details
Gene Symbol: HCG27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026791.1 | 2 | 752 | 136 | ||
GenBank mRNA | AK097442.1 | - | 2 | 752 | 136 | |
BC041465.2 | - | 1 | 631 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000361.hg.1, TC6_cox_hap2000058.hg.1, TC6_dbb_hap3000051.hg.1, TC6_mcf_hap5000045.hg.1, TC6_qbl_hap6000051.hg.1, TC6_ssto_hap7000045.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157671 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 30305 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 807961 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 652304 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 905932 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 238145 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 191226 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 51176 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 325645 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 69494 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 61784 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P21246 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 3032 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44248 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_14_P109495 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P21246 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P21246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642856 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642868 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642868 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15799838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24834450 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642856 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04642860 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P21246 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P109495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642854 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642886 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642862 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642868 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642856 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642865 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642858 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642864 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15799839 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642892 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642852 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642883 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642889 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642874 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642881 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04642877 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17496940 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P31720695 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P109495 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8179236 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8177953 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8118094 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8118096 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8177954 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8118095 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8118097 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8177952 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8179237 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8177951 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8179235 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8118093 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1559050_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000842.hg.1 | TC6_cox_hap2000058.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000842.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000696.hg.1 | TC6_mcf_hap5000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000696.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000837.hg.1 | TC6_qbl_hap6000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000837.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000823.hg.1 | TC6_dbb_hap3000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000823.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000841.hg.1 | TC6_cox_hap2000058.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000841.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003440.hg.1 | TC06000361.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003440.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003440.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003441.hg.1 | TC06000361.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003441.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003441.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000633.hg.1 | TC6_ssto_hap7000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000633.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000414008.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000414008.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000415276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000415276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000634.hg.1 | TC6_ssto_hap7000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000634.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000824.hg.1 | TC6_dbb_hap3000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000824.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000697.hg.1 | TC6_mcf_hap5000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000697.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000838.hg.1 | TC6_qbl_hap6000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000838.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151615.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151615.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151615.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151615.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151615.hg.1 | TC0600007568.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000414008.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000414008.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000415276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000415276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007568.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | ENST00000383331.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | ENST00000424675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | NR_026791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151614.hg.1 | TC0600007568.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1559050_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559050_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559050_at6 | - | - |
Human Genome U95 Set | 51293_at | - | - |
Human Genome U95 Set | 51293_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031273 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031273 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 234135 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1746436 | BC041465.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1746436 | AK097442.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1746436 | BC041465.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1746436 | NR_026791.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1746436 | AK097442.1 | human_transcriptome |
