Assay Details
| Gene Symbol: TSPY26P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002781.1 | 1 | 940 | 88 | ||
| GenBank mRNA | BC101556.1 | - | 1 | 940 | 88 | |
| BC101558.1 | - | 1 | 940 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000761.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P408341 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P408341 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 4468 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 34298 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P408341 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P408341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133007 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132993 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132993 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133007 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31780156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11132994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11133002 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P408341 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P102816 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P5994 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P408341 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P34683926 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132952 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132987 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132970 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132960 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132984 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132974 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132988 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133010 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132968 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133012 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132996 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132961 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132957 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133017 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17186829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132953 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132997 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132981 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133020 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133003 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132998 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132972 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132991 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133000 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132964 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133002 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133040 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133019 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11133007 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11132993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002781_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8065605 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8065604 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8065606 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8065603 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 233617_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010139.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010139.hg.1 | TC20000761.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008844.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | TC2000008844.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000161664.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000476365.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | ENST00000565928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | NR_002781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008844.hg.1 | NR_002781.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 233617_at | - | - |
| Human Genome U133 Plus 2.0 Array | 233617_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002781_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233617_at1 | - | - |
| Human Genome U133 Set | 233617_at | - | - |
| Human Genome U133 Set | 233617_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017550 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017550 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 176417 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737360 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1737360 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737360 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2231478 | NR_002781.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231478 | BC101556.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231478 | BC101558.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2231478 | NR_002781.1 | human_transcriptome |

TSPY26P
Genomic Map