Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018104.hg.1 |
ENST00000334636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018104.hg.1 |
ENST00000530609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018104.hg.1 |
ENST00000532378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018104.hg.1 |
NM_024514.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018104.hg.1 |
TC11001444.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000334636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000525015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000530609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000532378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000532805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000534686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
NM_024514.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000334636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000525015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000530609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000532378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000532805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
ENST00000534686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
NM_024514.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100177209.hg.1 |
TC1100010185.hg.1 |
Clariom D TC |