Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001021.4 | NP_001012.1 | 4 - 5 | 443 | 93 | |
NR_111943.1 | 3 - 4 | 649 | 93 | |||
GenBank mRNA | AK311951.1 | - | 4 - 5 | 351 | 93 | |
BC009407.2 | - | 1 | 334 | 4334552 | 93 | |
BC019899.1 | - | 1 | 339 | 3882924 | 93 | |
BC020453.1 | - | 1 | 338 | 93 | ||
BC022370.1 | - | 4 - 5 | 356 | 4738331 | 93 | |
BC049824.1 | - | 1 | 331 | 6212543 | 93 | |
BC062715.1 | - | 4 - 5 | 441 | 4068842 | 93 | |
BC070222.1 | - | 1 | 352 | 6738383 | 93 | |
BC071928.1 | - | 1 | 342 | 5090053 | 93 | |
BE905890.1 | - | 3 - 4 | 319 | 93 | ||
BI597659.1 | - | 3 - 4 | 649 | 93 | ||
BM017772.1 | - | 4 - 5 | 445 | 93 | ||
M13932.1 | - | 4 - 5 | 352 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte homeostasis
ribosomal small subunit biogenesis
structural constituent of ribosome
poly(A) RNA binding
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte homeostasis
ribosomal small subunit biogenesis
structural constituent of ribosome
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 873401 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 146474 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P418418 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P418418 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P117721 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 24017 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 21783 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57131 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58191 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P418418 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P418418 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P418418 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P418418 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P117721 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P418418 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P117721 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P916705 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7990966 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990917 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990917 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990968 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990920 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990969 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990969 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990918 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990970 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990918 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990921 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990919 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990967 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990968 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990919 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990966 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990967 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7990920 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7990916 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7990965 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7990965 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7990916 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 201665_PM_x_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 211487_PM_x_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 212578_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11744601_x_at | - | - |
Affymetrix Human Genome U219 Array | 11744334_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11744601_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11744334_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 211487_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 201665_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 212578_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 211487_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 216348_at | - | - |
Human Genome U133 Plus 2.0 Array | 216348_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 201665_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 212578_x_at | - | - |
Human Genome U133A 2.0 Array | 211487_x_at | - | - |
Human Genome U133A 2.0 Array | 212578_x_at | - | - |
Human Genome U133A 2.0 Array | 201665_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 201665_x_at | - | - |
Human Genome U133A 2.0 Array | 211487_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 212578_PM_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001021_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw564104 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw512208 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw573412 | - | - |
Human Genome U95 Set | 34592_PM_at | - | - |
Human Genome U95 Set | 34592_at | - | - |
Human Genome U95 Set | 34593_PM_g_at | - | - |
Human Genome U95 Set | 34593_g_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211487_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 212578_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 201665_x_at8 | - | - |
Human Genome U133 Set | 201665_PM_x_at | - | - |
Human Genome U133 Set | 201665_x_at | - | - |
Human Genome U133 Set | 212578_x_at | - | - |
Human Genome U133 Set | 211487_x_at | - | - |
Human Genome U133 Set | 211487_PM_x_at | - | - |
Human Genome U133 Set | 212578_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052473 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020348 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020348 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052473 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052473 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052473 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020348 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020348 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148139 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 175867 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2207539 | NM_001199057.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207539 | BC022370.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207539 | BC062715.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207539 | NM_001021.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207533 | CR618357.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207533 | CR620738.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207533 | M13932.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1685653 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207539 | NM_001199057.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207539 | BC022370.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207539 | BC062715.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2207539 | NM_001021.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_14591913-S | NM_001199057.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_14591913-S | BC022370.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_14591913-S | BC062715.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_14591913-S | BC070222.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_14591913-S | NM_001021.3 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27020-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0006620112 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0006620112 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1685653 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1685653 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_27020 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27020 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207539 | NM_001199057.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207539 | BC022370.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207539 | BC062715.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207539 | NM_001021.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207539 | NM_001199057.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207539 | BC022370.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207539 | BC062715.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207539 | NM_001021.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2207533 | CR615408.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1685653 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2207533 | NM_001199057.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2207533 | CR618357.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2207533 | CR620738.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2207533 | M13932.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2207533 | CR615408.1 | human_transcriptome |
