Assay Details
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174894.2 | NP_777554.1 | 1 - 2 | 232 | 59 | |
GenBank mRNA | BC042161.1 | - | 1 - 2 | 178 | 4869377 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000135.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC042161.1,NM_174894.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19002117.hg.1 | ENST00000317378.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002117.hg.1 | ENST00000426877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002117.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002117.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002117.hg.1 | TC19000135.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | ENST00000317378.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | ENST00000426877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19002120.hg.1 | TC19000135.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | TC1900011658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199297.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | TC1900011658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199296.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | TC1900011658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199292.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | TC1900011658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900199290.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000317378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000426877.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000595985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | ENST00000596148.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042461.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_001042462.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011658.hg.1 | NM_174894.2 | Refseq |
