Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003452.hg.1 |
ENST00000376958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003452.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003452.hg.1 |
TC13000330.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007731.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152137.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152137.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152137.hg.1 |
ENST00000376958.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152137.hg.1 |
NM_180989.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152137.hg.1 |
TC1300007731.hg.1 |
Clariom D TC |