Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009728.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009728.hg.1 |
ENST00000377009.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009728.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009728.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009728.hg.1 |
TC20000710.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008669.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
TC2000008669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160693.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
NM_130794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
TC2000008669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
ENST00000377007.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
ENST00000377009.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
NM_080830.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160694.hg.1 |
NM_130794.1 |
Refseq |