Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
ENST00000298239.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
ENST00000358559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
ENST00000474170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003909.hg.1 |
TC10000337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
ENST00000298239.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
ENST00000358559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
ENST00000474170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003912.hg.1 |
TC10000337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171619.hg.1 |
TC1000010516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171623.hg.1 |
TC1000010516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |