Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000287773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
ENST00000454765.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_001161342.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007761.hg.1 |
NM_173490.7 |
Refseq |