Assay Details
Gene Symbol: RASSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182663.3 | NP_872604.1 | 4 - 5 | 1062 | 84 | |
NM_182665.3 | NP_872606.1 | 3 - 4 | 940 | 84 | ||
GenBank mRNA | AF445801.1 | - | 3 - 4 | 940 | 84 | |
AK074115.1 | - | 2 - 3 | 407 | 84 | ||
AK289861.1 | - | 4 - 5 | 1062 | 84 | ||
AK291268.1 | - | 4 - 5 | 1060 | 84 | ||
AK292039.1 | - | 4 - 5 | 798 | 84 | ||
AL832784.1 | - | 3 - 4 | 840 | 84 | ||
AY062002.1 | - | 4 - 5 | 1046 | 84 | ||
AY216268.1 | - | 3 - 4 | 933 | 84 | ||
AY261332.1 | - | 3 - 4 | 840 | 84 | ||
BC004270.1 | - | 4 - 5 | 1046 | 3613993 | 84 | |
BC007203.1 | - | 4 - 5 | 1046 | 2967057 | 84 | |
BC042651.1 | - | 3 - 4 | 920 | 4342962 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001724.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | TC01001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | ENST00000304534.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | ENST00000338603.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | ENST00000355294.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | ENST00000367118.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | ENST00000491368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01026904.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000577571.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000579436.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000579942.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000581503.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | ENST00000581888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | NM_182663.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | NM_182665.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100183653.hg.1 | TC0100011378.hg.1 | Clariom D TC |
