Assay Details
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003095.2 | NP_003086.1 | 1 - 2 | 139 | 154 | |
| GenBank mRNA | BC002505.2 | - | 1 - 2 | 132 | 3051263 | 154 |
| BU861031.1 | - | 1 - 2 | 142 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000742.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC002505.2,NM_003095.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12009387.hg.1 | ENST00000266735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009387.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009387.hg.1 | ENST00000553192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009387.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009387.hg.1 | TC12000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009394.hg.1 | ENST00000414761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009388.hg.1 | ENST00000266735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009388.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009388.hg.1 | ENST00000553192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009388.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009388.hg.1 | TC12000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009381.hg.1 | ENST00000266735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009381.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009381.hg.1 | TC12000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009384.hg.1 | ENST00000266735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009384.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009384.hg.1 | ENST00000553192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009384.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009384.hg.1 | TC12000742.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007424.hg.1 | ENST00000440710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009971.hg.1 | ENST00000564938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009971.hg.1 | ENST00000564938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011300.hg.1 | ENST00000433886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010076.hg.1 | ENST00000566222.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162476.hg.1 | TC1200008525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011300.hg.1 | ENST00000433886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010076.hg.1 | ENST00000566222.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009394.hg.1 | ENST00000414761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162473.hg.1 | TC1200008525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162474.hg.1 | TC1200008525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200162469.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162469.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162469.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162469.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162469.hg.1 | TC1200008525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007424.hg.1 | ENST00000440710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000266735.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000549580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000551316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000552085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | ENST00000553192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008525.hg.1 | NM_003095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009971.hg.1 | ENST00000564938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009971.hg.1 | ENST00000564938.1 | Ensembl |

SNRPF
Genomic Map