Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001551.2 | NP_001542.1 | 1 - 2 | 71 | 97 | |
| GenBank mRNA | BC004137.2 | - | 1 - 2 | 71 | 2962967 | 97 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000370.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC004137.2,NM_001551.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
signal transduction
negative regulation of stress-activated MAPK cascade
regulation of protein phosphatase type 2A activity
response to tumor necrosis factor
positive regulation of dephosphorylation
negative regulation of protein dephosphorylation
B cell activation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription from RNA polymerase II promoter
regulation of microtubule-based movement
response to interleukin-1
negative regulation of apoptotic signaling pathway
protein binding
protein phosphatase type 2A regulator activity
protein domain specific binding
mitogen-activated protein kinase kinase binding
protein complex binding
protein phosphatase 2A binding
negative regulation of transcription from RNA polymerase II promoter
signal transduction
negative regulation of stress-activated MAPK cascade
regulation of protein phosphatase type 2A activity
response to tumor necrosis factor
positive regulation of dephosphorylation
negative regulation of protein dephosphorylation
B cell activation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription from RNA polymerase II promoter
regulation of microtubule-based movement
response to interleukin-1
negative regulation of apoptotic signaling pathway
protein binding
protein phosphatase type 2A regulator activity
protein domain specific binding
mitogen-activated protein kinase kinase binding
protein complex binding
protein phosphatase 2A binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006886.hg.1 | ENST00000554337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006886.hg.1 | ENST00000554337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006886.hg.1 | NR_002937.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006886.hg.1 | NR_002937.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010653.hg.1 | ENST00000447449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007141.hg.1 | ENST00000507524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004770.hg.1 | TC0X000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004770.hg.1 | ENST00000342206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004770.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004770.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011266.hg.1 | ENST00000583666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004768.hg.1 | TC0X000370.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004768.hg.1 | ENST00000342206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004768.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004768.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151544.hg.1 | TC0X00007529.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007529.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007141.hg.1 | ENST00000507524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006886.hg.1 | ENST00000554337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006886.hg.1 | ENST00000554337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006886.hg.1 | NR_002937.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006886.hg.1 | NR_002937.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011266.hg.1 | ENST00000583666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151542.hg.1 | TC0X00007529.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | ENST00000342206.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | ENST00000356413.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | NM_001551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151545.hg.1 | TC0X00007529.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010653.hg.1 | ENST00000447449.1 | Ensembl |

IGBP1
Genomic Map