Assay Details
| Gene Symbol: NFYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006166.3 | NP_006157.1 | 1 - 2 | 125 | 147 | |
| GenBank mRNA | BC005316.1 | - | 1 - 2 | 74 | 3680937 | 147 |
| BC005317.1 | - | 1 - 2 | 80 | 3997299 | 147 | |
| BC007035.1 | - | 1 - 2 | 52 | 3996244 | 147 | |
| BG776259.1 | - | 1 - 2 | 126 | 147 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001906.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC005316.1,BC005317.1,BC007035.1,NM_006166.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein complex binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
repressing transcription factor binding
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein complex binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
repressing transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024881.hg.1 | ENST00000240055.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024881.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024881.hg.1 | TC12001906.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024876.hg.1 | ENST00000240055.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024876.hg.1 | ENST00000551727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024876.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024876.hg.1 | TC12001906.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024878.hg.1 | ENST00000240055.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024878.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12024878.hg.1 | TC12001906.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011755.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | ENST00000551727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191178.hg.1 | TC1200011755.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000240055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000550881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | ENST00000551446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | NM_006166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191180.hg.1 | TC1200011755.hg.1 | Clariom D TC |

NFYB
Genomic Map