Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007080.2 | NP_009011.1 | 1 - 2 | 127 | 148 | |
GenBank mRNA | BC016026.1 | - | 1 - 2 | 81 | 3910698 | 148 |
DB465201.1 | - | 1 - 2 | 127 | 149 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000717.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC016026.1,NM_007080.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | TC04000717.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | ENST00000296581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | ENST00000502781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | ENST00000515311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010196.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007580.hg.1 | ENST00000552208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | TC04000717.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | ENST00000296581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | ENST00000502781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | ENST00000515311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010191.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007680.hg.1 | ENST00000604976.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400161315.hg.1 | TC0400008922.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007680.hg.1 | ENST00000604976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000507449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008922.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000296581.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000502781.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000503982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000504181.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000510331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | ENST00000515311.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | NM_007080.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400161309.hg.1 | TC0400008922.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200007580.hg.1 | ENST00000552208.1 | Ensembl |
