Assay Details
| Gene Symbol: STK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC005231.1 | - | 2 | 654 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000372806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000474717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000480745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000487587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000488618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | ENST00000499879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007476.hg.1 | NM_006282.4 | Refseq |

STK4
Genomic Map