Assay Details
| Gene Symbol: GUSBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003675.2 | 1 | 738 | 162 | ||
| GenBank mRNA | BC025996.2 | - | 1 | 738 | 162 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 382579 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 66463 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P406591 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 42679 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37085 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P406591 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P406591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653927 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653919 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653880 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653882 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653885 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653927 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653882 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653880 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653919 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653885 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23653903 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P406591 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P406591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493255 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493289 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15621580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15621581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493318 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493258 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493292 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03493280 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003675_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8097627 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8097626 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 240063_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 240063_at | - | - |
| Human Genome U133 Plus 2.0 Array | 240063_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 240063_at8 | - | - |
| Human Genome U95 Set | 77754_PM_at | - | - |
| Human Genome U95 Set | 77754_at | - | - |
| Human Genome U133 Set | 240063_at | - | - |
| Human Genome U133 Set | 240063_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026096 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026096 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 224343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1693523 | BC025996.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1693523 | NR_003675.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1693523 | BC025996.2 | human_transcriptome |

GUSBP5
Genomic Map