Assay Details
Gene Symbol: LOC403312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301851.1 | NP_001288780.1 | 2 | 2809 | 114 | |
GenBank mRNA | BC036197.1 | - | 3 | 2780 | 5273873 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524403 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524401 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28524404 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242947 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 189472 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 334657 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P170970 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 40933 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8933 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P33076971 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3230985 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P170970 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02540525 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P170970 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02540525 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P800497 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02540525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7944973 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7944977 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7944974 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7944975 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7944976 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1555002_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170662.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170662.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170662.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170662.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170662.hg.1 | TC1100009418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009418.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170663.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170663.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170663.hg.1 | ENST00000527818.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170663.hg.1 | NM_001301851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170663.hg.1 | TC1100009418.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1555002_at | - | - |
Human Genome U133 Plus 2.0 Array | 1555002_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555002_at7 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1708535 | BC036197.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708535 | BC036197.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1708535 | BC036197.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708535 | BC036197.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708535 | BC036197.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708535 | BC036197.1 | human_transcriptome |
