Assay Details
| Gene Symbol: ACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036253.1 | - | 1 | 146 | 5268748 | 131 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC036253.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200154045.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154045.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154045.hg.1 | TC0200007835.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |

ACTR2
Genomic Map