Assay Details
| Gene Symbol: NUDT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304963.1 | NP_001291892.1 | 2 | 1015 | 151 | |
| NM_153183.3 | NP_694853.1 | 3 | 1078 | 151 | ||
| GenBank mRNA | AF469196.1 | - | 3 | 1080 | 151 | |
| AK291952.1 | - | 2 | 995 | 151 | ||
| BC049383.1 | - | 2 | 936 | 5725355 | 151 | |
| BC050700.1 | - | 2 | 994 | 5723622 | 151 | |
| CN399689.1 | - | 2 | 684 | 151 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUDT10
Genomic Map