Assay Details
| Gene Symbol: GAMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138924.2 | NP_620279.1 | 5 | 768 | 134 | |
| GenBank mRNA | AF086508.1 | - | 3 | 353 | 134 | |
| AK289465.1 | - | 5 | 742 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
creatine metabolic process
creatine biosynthetic process
muscle contraction
spermatogenesis
organ morphogenesis
methylation
regulation of multicellular organism growth
S-adenosylhomocysteine metabolic process
S-adenosylmethionine metabolic process
embryonic liver development
methyltransferase activity
guanidinoacetate N-methyltransferase activity
protein homodimerization activity
creatine metabolic process
creatine biosynthetic process
muscle contraction
spermatogenesis
organ morphogenesis
methylation
regulation of multicellular organism growth
S-adenosylhomocysteine metabolic process
S-adenosylmethionine metabolic process
embryonic liver development
methyltransferase activity
guanidinoacetate N-methyltransferase activity
protein homodimerization activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172749.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172749.hg.1 | TC1900009155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172749.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172750.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172750.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172750.hg.1 | TC1900009155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172750.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172750.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172754.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172754.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172754.hg.1 | TC1900009155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172754.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172754.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | TC1900009155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172752.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000252288.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_000156.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009155.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | NM_138924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | TC1900009155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | ENST00000447102.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | ENST00000591788.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172753.hg.1 | NM_138924.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 225790 | - | - |

GAMT
Genomic Map