Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145165.1 | NP_001138637.1 | 2 - 3 | 722 | 84 | |
| NM_031304.4 | NP_112594.1 | 2 - 3 | 433 | 84 | ||
| XM_011528341.2 | XP_011526643.1 | 2 - 3 | 430 | 84 | ||
| GenBank mRNA | BC002817.2 | - | 2 - 3 | 433 | 3635058 | 84 |
| BC009863.2 | - | 1 - 2 | 322 | 3938535 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001043.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014215.hg.1 | ENST00000250937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014215.hg.1 | ENST00000427575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014215.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014215.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014215.hg.1 | TC19001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014220.hg.1 | ENST00000427575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014220.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014220.hg.1 | TC19001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174019.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174027.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174027.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174027.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174027.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174027.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174026.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | NM_031304.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | TC1900009289.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000250937.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000427575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000586906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000587122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | ENST00000592858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | NM_001145165.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174017.hg.1 | NM_031304.4 | Refseq |

DOHH
Genomic Map