Assay Details
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018002.3 | NP_060472.2 | 11 - 12 | 2291 | 86 | |
NM_181354.4 | NP_851999.2 | 9 - 10 | 2121 | 86 | ||
XM_006716595.2 | XP_006716658.1 | 10 - 11 | 2060 | 86 | ||
XM_017013594.1 | XP_016869083.1 | 9 - 10 | 1995 | 86 | ||
XM_017013599.1 | XP_016869088.1 | 8 - 9 | 1905 | 86 | ||
XM_017013600.1 | XP_016869089.1 | 8 - 9 | 1762 | 86 | ||
GenBank mRNA | AK000987.1 | - | 2 - 3 | 262 | 86 | |
AK292899.1 | - | 9 - 10 | 2073 | 86 | ||
AL833193.1 | - | 8 - 9 | 1702 | 86 | ||
BC032710.2 | - | 9 - 10 | 2063 | 5496492 | 86 | |
BC035484.1 | - | 9 - 10 | 2075 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
