Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084392.1 | NP_001077861.1 | 2 - 3 | 364 | 93 | |
| NM_001355.3 | NP_001346.1 | 3 - 4 | 415 | 93 | ||
| GenBank mRNA | AK292044.1 | - | 2 - 3 | 308 | 93 | |
| BC005971.1 | - | 2 - 3 | 312 | 4082777 | 93 | |
| BC015508.2 | - | 2 - 3 | 333 | 3902943 | 93 | |
| BT006970.1 | - | 2 - 3 | 287 | 93 | ||
| CA414359.1 | - | 1 - 2 | 93 | |||
| CR456431.1 | - | 2 - 3 | 310 | 93 | ||
| U49785.1 | - | 3 - 4 | 413 | 93 | ||
| U84143.1 | - | 2 - 3 | 301 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001486.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22010241.hg.1 | TC22001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010241.hg.1 | ENST00000398344.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010241.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | TC22001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | ENST00000350608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | ENST00000398344.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | ENST00000404092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010229.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158766.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158776.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158767.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158774.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158775.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | TC2200008203.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158779.hg.1 | NM_001084392.1 | Refseq |

DDT
Genomic Map