Assay Details
| Gene Symbol: FLJ32154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056716.1 | - | 1 - 2 | 297 | 105 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P384551 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P384551 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 43340 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P133602 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P133602 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49840 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P384551 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P133602 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P133602 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 70150 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 91459 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P133602 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8067545 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8067544 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8067543 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 235933_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009765.hg.1 | ENST00000370520.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009765.hg.1 | ENST00000370520.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009765.hg.1 | ENST00000370520.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009765.hg.1 | ENST00000370520.3 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 235933_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235933_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235933_at8 | - | - |
| Human Genome U95 Set | 84750_PM_at | - | - |
| Human Genome U95 Set | 84750_at | - | - |
| Human Genome U133 Set | 235933_at | - | - |
| Human Genome U133 Set | 235933_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 221965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1671331 | AK056716.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1671331 | AK056716.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1671331 | AK056716.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1671331 | AK056716.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1671331 | AK056716.1 | human_transcriptome |

FLJ32154
Genomic Map