Assay Details
| Gene Symbol: IRF2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182972.2 | NP_892017.2 | 1 - 2 | 1078 | 77 | |
| GenBank mRNA | AY278023.1 | - | 1 - 2 | 1078 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003985.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | TC01003985.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | ENST00000366610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060308.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060306.hg.1 | TC01003985.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060306.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060306.hg.1 | ENST00000366610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060306.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060306.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060307.hg.1 | TC01003985.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060307.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060307.hg.1 | ENST00000366610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060307.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060307.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234217.hg.1 | TC0100017793.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017793.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | ENST00000491430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234218.hg.1 | TC0100017793.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | ENST00000366609.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | ENST00000366610.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | NM_001077397.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | NM_182972.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234215.hg.1 | TC0100017793.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 191794 | - | - |

IRF2BP2
Genomic Map