Assay Details
| Gene Symbol: PRSS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135086.1 | NP_001128558.1 | 4 - 5 | 744 | 70 | |
| XM_011522477.2 | XP_011520779.1 | 6 - 7 | 1362 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000085.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 48835 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 122077 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 464978 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 133909 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416634 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1857 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1856 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5095 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P40537099 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P40537099 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416634 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3416634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30401756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16844239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30401742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16844227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16844239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30401756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30401742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16844227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09891639 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891658 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891671 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891665 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891632 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891647 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891639 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891629 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891626 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891660 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16844228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891640 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891641 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891636 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16844232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891649 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16844225 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891638 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16844239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891643 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891627 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09891645 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P40537099 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03103759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7992719 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7992717 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7992718 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7992720 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7992721 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7992716 | - | - |
| Affymetrix Human Genome U219 Array | 11743215_at | - | - |
| Affymetrix Human Genome U219 Array | 11743215_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001597.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001597.hg.1 | TC16000085.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006635.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | ENST00000399677.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | ENST00000421271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | NM_001135086.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600147020.hg.1 | TC1600006635.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054210 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054210 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 203961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1667596 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1667596 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1651230 | NM_001135086.1 | human_transcriptome |

PRSS41
Genomic Map