Assay Details
Gene Symbol: LOC157740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ291676.1 | - | 1 | 1434 | 136 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | AK024419.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | AK074043.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | AK074548.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | AK090438.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | AK289533.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC000158.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC009256.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC011667.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC020512.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC033749.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC035961.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC050605.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC071644.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC071876.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | BC094816.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | CR593932.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | CR594563.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | CR607418.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | CR615865.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | CR624450.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 157732 | NM_031213.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 139658 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 152685 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 19600 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 138181 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 41995 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 37102 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 4163 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 24253 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 21932 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 828961 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 385777 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 638533 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 736896 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 560635 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187010 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 58079 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 92120 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P200237 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P200237 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 33988 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P109412 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P109412 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48561 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P200237 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P200237 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P109412 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P109412 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P109412 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P200237 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 81342 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 72002 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P109412 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P200237 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P109412 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | XR_042030_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XR_042031_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XR_042029_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8149321 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8149320 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 239877_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009595.hg.1 | ENST00000622929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009595.hg.1 | ENST00000622929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000221086.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000526292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000528389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | ENST00000530200.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006738.hg.1 | NM_015458.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 239877_at | - | - |
Human Genome U133 Plus 2.0 Array | 239877_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239877_at5 | - | - |
Human Genome U95 Set | 74249_at | - | - |
Human Genome U95 Set | 74249_PM_at | - | - |
Human Genome U133 Set | 239877_at | - | - |
Human Genome U133 Set | 239877_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 228213 | - | - |
