Assay Details
| Gene Symbol: DNAJC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012339.2 | NP_001012339.2 | 5 - 6 | 963 | 65 | |
| NM_194283.3 | NP_919259.3 | 5 - 6 | 963 | 65 | ||
| XM_005248249.4 | XP_005248306.2 | 5 - 6 | 1016 | 65 | ||
| XM_005248250.3 | XP_005248307.2 | 5 - 6 | 1016 | 65 | ||
| XM_011513965.2 | XP_011512267.2 | 5 - 6 | 1016 | 65 | ||
| XM_011513966.2 | XP_011512268.2 | 5 - 6 | 1016 | 65 | ||
| GenBank mRNA | AK127749.1 | - | 2 - 3 | 393 | 65 | |
| AY177688.1 | - | 5 - 6 | 972 | 65 | ||
| AY258422.1 | - | 5 - 6 | 966 | 65 | ||
| BC050472.1 | - | 1 - 2 | 263 | 65 | ||
| BC107577.1 | - | 2 - 3 | 363 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000155.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | TC05000155.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | ENST00000303525.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | ENST00000342382.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001689.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148127.hg.1 | TC0500007123.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000342382.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000382021.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000506762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000509626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000512136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | ENST00000514237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_001012339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007123.hg.1 | NM_194283.3 | Refseq |

DNAJC21
Genomic Map