Assay Details
| Gene Symbol: WFDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF330260.1 | - | 1 - 2 | 90 | 179 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |

WFDC2
Genomic Map