Assay Details
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC022294.1 | - | 4 | 533 | 4716526 | 220 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21419909 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3300495 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P359630 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P197825 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2275 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2276 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44272 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P220750 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P197825 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00320333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21419876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21419876 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P197825 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P359630 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P197822 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8044152 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043371 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043373 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043370 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053608 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053604 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053607 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043369 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8044153 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053606 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053605 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8044150 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8053609 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8043372 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8043367 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8044149 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8053602 | - | - |
Affymetrix Human Genome U219 Array | 11739531_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11761775_s_at | - | - |
Affymetrix Human Genome U219 Array | 11739530_at | - | - |
Affymetrix Human Genome U219 Array | 11739531_x_at | - | - |
Affymetrix Human Genome U219 Array | 11761775_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11739530_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214415_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 205871_at | - | - |
Human Genome U133 Plus 2.0 Array | 230120_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 214415_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558603_at | - | - |
Human Genome U133 Plus 2.0 Array | 205871_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 230120_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558603_PM_at | - | - |
Human Genome U133A 2.0 Array | 214415_PM_at | - | - |
Human Genome U133A 2.0 Array | 214415_at | - | - |
Human Genome U133A 2.0 Array | 205871_at | - | - |
Human Genome U133A 2.0 Array | 205871_PM_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw502395 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 205871_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230120_s_at4 | - | - |
Human Genome U95 Set | 49814_at | - | - |
Human Genome U95 Set | 58588_PM_at | - | - |
Human Genome U95 Set | 49814_PM_at | - | - |
Human Genome U95 Set | 58588_at | - | - |
Human Genome U133 Set | 205871_PM_at | - | - |
Human Genome U133 Set | 214415_at | - | - |
Human Genome U133 Set | 205871_at | - | - |
Human Genome U133 Set | 214415_PM_at | - | - |
Human Genome U133 Set | 230120_s_at | - | - |
Human Genome U133 Set | 230120_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013972 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014166 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014166 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3238364 | NM_001032392.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3238364 | NM_002665.4 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698587 | BC070296.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698587 | BC107767.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698587 | BC109248.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698587 | BC005379.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698587 | BC022294.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238364 | NM_001032392.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3238364 | NM_002665.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238364 | NM_001032392.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238364 | NM_002665.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238364 | NM_001032392.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238364 | NM_002665.4 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20070169-S | NM_001024457.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20070169-S | CR749330.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20070169-S | NR_003506.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20070169-S | U67178.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20070169-S | BC070296.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_3852-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001190333 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001190333 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1676779 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1698587 | BC070296.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1698587 | BC107767.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1698587 | BC109248.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1698587 | BC005379.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1698587 | BC022294.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1751811 | NM_001032392.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1751811 | NM_002665.4 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698587 | BC070296.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698587 | BC107767.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698587 | BC109248.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698587 | BC005379.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698587 | BC022294.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 50746 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_3852 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698587 | BC070296.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698587 | BC107767.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698587 | BC109248.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698587 | BC005379.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698587 | BC022294.1 | human_transcriptome |
