Assay Details
| Gene Symbol: RPS3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006.4 | NP_000997.1 | 1 - 2 | 145 | 97 | |
| NM_001267699.1 | NP_001254628.1 | 1 - 2 | 145 | 97 | ||
| GenBank mRNA | AK129825.1 | - | 1 - 2 | 88 | 97 | |
| AK291075.1 | - | 1 - 2 | 89 | 97 | ||
| AK308947.1 | - | 1 - 2 | 90 | 97 | ||
| AK311788.1 | - | 1 - 2 | 88 | 97 | ||
| BC000204.1 | - | 1 | 77 | 3350750 | 97 | |
| BC001708.1 | - | 1 - 2 | 107 | 3544072 | 97 | |
| BC004981.1 | - | 1 - 2 | 77 | 2906117 | 97 | |
| BC006298.2 | - | 1 | 70 | 4079651 | 97 | |
| BC009219.2 | - | 1 | 77 | 3927639 | 97 | |
| BC009404.2 | - | 1 | 79 | 4308955 | 97 | |
| BC017123.2 | - | 1 | 77 | 3922810 | 97 | |
| BC018954.2 | - | 1 | 79 | 97 | ||
| BC019072.2 | - | 1 | 128 | 5020581 | 97 | |
| BC030161.2 | - | 1 | 128 | 5020904 | 97 | |
| BC066926.1 | - | 1 | 89 | 4826149 | 97 | |
| BC070211.1 | - | 1 | 90 | 5809691 | 97 | |
| BC071916.1 | - | 1 | 77 | 6285242 | 97 | |
| BQ055367.1 | - | 1 - 2 | 116 | 97 | ||
| CX754149.1 | - | 1 | 145 | 97 | ||
| L13802.1 | - | 1 - 2 | 71 | 97 | ||
| M77234.1 | - | 1 - 2 | 82 | 97 | ||
| M84711.1 | - | 1 - 2 | 101 | 97 | ||
More Information
Set Membership: |
|

RPS3A
Genomic Map