Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000264.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000264.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000264.hg.1 |
TC11000019.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006473.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144577.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144577.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144577.hg.1 |
ENST00000270115.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144577.hg.1 |
NM_198075.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144577.hg.1 |
TC1100006473.hg.1 |
Clariom D TC |