Assay Details
| Gene Symbol: ESCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832041.1 | - | 8 - 9 | 3038 | 147 | |
| AL834200.1 | - | 5 - 6 | 1594 | 147 | ||
| BC036943.1 | - | 1 - 2 | 131 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC036943.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000269214.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000383276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000580101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | ENST00000622333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008231.hg.1 | NM_052911.2 | Refseq |

ESCO1
Genomic Map