Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007662.hg.1 |
ENST00000449661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004434.hg.1 |
TC09000415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004434.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004434.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004435.hg.1 |
TC09000415.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004435.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004435.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153014.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153014.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153014.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153014.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153014.hg.1 |
TC0900007863.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153015.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153015.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153015.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153015.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153015.hg.1 |
TC0900007863.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007863.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007662.hg.1 |
ENST00000449661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153017.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153017.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153017.hg.1 |
ENST00000314355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153017.hg.1 |
NM_001827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153017.hg.1 |
TC0900007863.hg.1 |
Clariom D TC |