Assay Details
Gene Symbol: RPL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021029.5 | NP_066357.2 | 4 | 371 | 160 | |
GenBank mRNA | AY927233.1 | - | 1 | 234 | 160 | |
BC001781.1 | - | 1 | 257 | 3353669 | 160 | |
BC031015.1 | - | 1 | 249 | 4659712 | 160 | |
BC062219.1 | - | 4 | 282 | 6726961 | 160 | |
BC070204.1 | - | 1 | 246 | 4774945 | 160 | |
BF211310.1 | - | 4 | 302 | 160 | ||
BG504049.1 | - | 4 | 250 | 160 | ||
CR542198.1 | - | 1 | 217 | 160 |
Gene Symbol: RPL36A-HNRNPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199973.1 | NP_001186902.1 | 4 | 371 | 160 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X002317.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700008437.hg.1 | ENST00000578177.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006213.hg.1 | TC0X002317.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006213.hg.1 | ENST00000427805.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006213.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006213.hg.1 | ENST00000553110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006213.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008285.hg.1 | ENST00000523014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009427.hg.1 | ENST00000605745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013243.hg.1 | ENST00000428733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177728.hg.1 | ENST00000409170.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177728.hg.1 | NM_001199973.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177728.hg.1 | ENST00000409170.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177728.hg.1 | NM_001199973.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177728.hg.1 | TC0X00011310.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465340.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000491851.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_001199972.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011308.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000372849.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000392994.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000427805.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000465744.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000471855.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000489407.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000553110.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | ENST00000614077.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | NM_021029.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177704.hg.1 | TC0X00011308.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008285.hg.1 | ENST00000523014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013243.hg.1 | ENST00000428733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008437.hg.1 | ENST00000578177.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | ENST00000409170.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | ENST00000409338.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | NM_001199973.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | NM_001199974.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | ENST00000409170.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | ENST00000409338.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | NM_001199973.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | NM_001199974.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00177727.hg.1 | TC0X00011310.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500009427.hg.1 | ENST00000605745.1 | Ensembl |
