| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000309451.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000410036.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000410036.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000410036.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000488586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000488586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000491265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
ENST00000579560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NM_001085365.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016732.hg.1 |
NR_039945.1 |
Refseq |