Assay Details
| Gene Symbol: TMPRSS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182973.1 | NP_892018.1 | 2 - 3 | 277 | 123 | |
| GenBank mRNA | AJ488946.1 | - | 2 - 3 | 277 | 123 | |
| AJ488947.1 | - | 2 - 3 | 277 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000054.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000852.hg.1 | ENST00000332578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000852.hg.1 | ENST00000395264.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000852.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19000852.hg.1 | TC19000054.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000587863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000592650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006586.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | NM_182973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | TC1900006586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000332578.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000395264.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | ENST00000613480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900145920.hg.1 | NM_182973.2 | Refseq |

TMPRSS9
Genomic Map