Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175776.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175776.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175776.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175776.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175776.hg.1 |
TC0100010320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175775.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175775.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175775.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175775.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175775.hg.1 |
TC0100010320.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010320.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175777.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175777.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175777.hg.1 |
ENST00000356006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175777.hg.1 |
NR_110695.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175777.hg.1 |
TC0100010320.hg.1 |
Clariom D TC |