| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008844.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008844.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008844.hg.1 |
TC13000779.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008843.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008843.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008843.hg.1 |
TC13000779.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008845.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008845.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008845.hg.1 |
TC13000779.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009456.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163319.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163319.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163319.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163319.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163319.hg.1 |
TC1300009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163320.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163320.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163320.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163320.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163320.hg.1 |
TC1300009456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300163321.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163321.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163321.hg.1 |
ENST00000419272.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300163321.hg.1 |
NR_027039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300163321.hg.1 |
TC1300009456.hg.1 |
Clariom D TC |