Assay Details
| Gene Symbol: STX17-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038853.1 | 2 - 3 | 174 | 81 | ||
| GenBank mRNA | BC030123.1 | - | 2 - 3 | 180 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001414.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16247724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848661 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27191995 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848609 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27191997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192052 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27191999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27191996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192020 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192039 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192018 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192049 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848670 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192038 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848614 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16247723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848663 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16247730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27192063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06848646 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 1663 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 523578 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 898491 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 210304 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 636396 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 870363 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 797469 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 238408 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 286785 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 163784 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 391159 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 33927 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 55672 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P177955 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 19028 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24068 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18711363 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P38812245 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P38812245 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02142411 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P177955 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P177955 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P38812245 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P38812270 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P38812245 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 139391 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 101268 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18711365 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38812276 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38812245 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18711362 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18711353 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02142395 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P26668599 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18711357 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18711351 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38812270 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P177959 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38812245 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18711353 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38812270 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1558689_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1558688_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017645.hg.1 | TC09001414.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017645.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017645.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017647.hg.1 | TC09001414.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017647.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017647.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017646.hg.1 | TC09001414.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017646.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017646.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175075.hg.1 | TC0900011010.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175072.hg.1 | TC0900011010.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175073.hg.1 | TC0900011010.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | ENST00000529965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011010.hg.1 | NR_038853.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1558689_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1558688_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1558689_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1558688_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558689_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1558688_at11 | - | - |
| Human Genome U95 Set | 84417_PM_at | - | - |
| Human Genome U95 Set | 84417_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037809 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 290347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1702496 | NR_038853.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1702496 | BC030123.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1702496 | NR_038853.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1702496 | BC030123.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1702496 | NR_038853.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1702496 | BC030123.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1702496 | NR_038853.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1702496 | BC030123.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1702496 | NR_038853.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1702496 | BC030123.1 | human_transcriptome |

STX17-AS1
Genomic Map