Assay Details
| Gene Symbol: LINC00173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027346.1 | 2 - 3 | 273 | 66 | ||
| GenBank mRNA | BC038547.1 | - | 2 - 3 | 273 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000912.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08636855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08636854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08636852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08636853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08636850 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 673048 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3331511 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5131 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5130 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5128 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5129 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21830 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3331511 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3331511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08636788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29072020 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636745 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636813 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636852 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16595024 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636846 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636824 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636848 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636793 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636855 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636838 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636843 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636822 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636849 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636832 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636763 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636757 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636800 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636810 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636790 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636798 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16595022 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636777 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636775 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636837 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16595030 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636815 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636779 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636804 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636808 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636827 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636819 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636792 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08636797 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19694722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7959019 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7959017 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7959022 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7959020 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7959021 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7959018 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7959016 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1563369_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 237591_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011912.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011912.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011912.hg.1 | TC12000912.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000489452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | ENST00000489452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201721.hg.1 | TC1200012711.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000470091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000477702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000480237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000489452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | ENST00000489452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012711.hg.1 | NR_027346.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007503 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049079 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049080 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049080 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049079 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 638716 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3311190 | BC038547.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3311190 | NR_027346.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3311165 | BC121822.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3311165 | BC122872.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3311165 | NR_027345.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3311165 | AK124947.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311190 | BC038547.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311190 | NR_027346.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311165 | BC121822.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311165 | BC122872.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311165 | NR_027345.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311165 | AK124947.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311190 | BC038547.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311190 | NR_027346.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311190 | BC038547.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311190 | NR_027346.1 | human_transcriptome |

LINC00173
Genomic Map