Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146910.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146910.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146910.hg.1 |
TC1400006743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400146909.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400146909.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146909.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400146909.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146909.hg.1 |
TC1400006743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400146908.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146908.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400146908.hg.1 |
TC1400006743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
ENST00000555109.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006743.hg.1 |
NR_110032.1 |
Refseq |