Assay Details
| Gene Symbol: HNRNPUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079559.2 | NP_001073027.1 | 3 - 4 | 976 | 71 | |
| GenBank mRNA | BC029105.1 | - | 3 - 4 | 1063 | 71 | |
| DQ470474.1 | - | 3 - 4 | 940 | 71 | ||
| Gene Symbol: HNRNPUL2-BSCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037946.1 | 3 - 4 | 975 | 71 | ||
| GenBank mRNA | AK122942.1 | - | 3 - 4 | 975 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003488.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK122942.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021979.hg.1 | TC11003488.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021979.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021979.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204425.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204425.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204425.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204425.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204425.hg.1 | TC1100013175.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204443.hg.1 | TC1100013176.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |

HNRNPUL2
Genomic Map