Assay Details
Gene Symbol: HNRNPUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079559.2 | NP_001073027.1 | 6 - 7 | 1320 | 111 | |
GenBank mRNA | BC029105.1 | - | 6 - 7 | 1407 | 111 | |
DQ470474.1 | - | 6 - 7 | 1284 | 111 |
Gene Symbol: HNRNPUL2-BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037946.1 | 6 - 7 | 1319 | 111 | ||
GenBank mRNA | AK122942.1 | - | 6 - 7 | 1319 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11003488.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK122942.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11021976.hg.1 | TC11003488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11021976.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11021976.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204439.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204439.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204439.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204439.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204439.hg.1 | TC1100013176.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100204422.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204422.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204422.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204422.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204422.hg.1 | TC1100013175.hg.1 | Clariom D TC |
