Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001277.hg.1 |
TC07000075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001277.hg.1 |
ENST00000223122.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001277.hg.1 |
ENST00000436587.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001277.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000402468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000419721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000429911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000467716.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
ENST00000476068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006662.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146096.hg.1 |
TC0700006662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
ENST00000223122.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
ENST00000436587.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
NM_020156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146094.hg.1 |
TC0700006662.hg.1 |
Clariom D TC |